  • Search Results
  • Role of Acyl Chain Composition of Phosphatidylcholine in Tafazzin-Mediated Remodeling of Cardiolipin in Liposomes.
Role of Acyl Chain Composition of Phosphatidylcholine in Tafazzin-Mediated Remodeling of Cardiolipin in Liposomes.
Biochemistry (2017-11-02)
Masato Abe, Yoshiki Sawada, Shinpei Uno, Shuhei Chigasaki, Masahide Oku, Yasuyoshi Sakai, Hideto Miyoshi
ABSTRACT
Remodeling of the acyl chain compositions of cardiolipin (CL) species by the transacylase tafazzin is an important process for maintaining optimal mitochondrial functions. The results of mechanistic studies on the tafazzin-mediated transacylation from phosphatidylcholine (PC) to monolyso-CL (MLCL) in artificial lipid membranes are controversial. The present study investigated the role of the acyl chain composition of PC in the Saccharomyces cerevisiae tafazzin-mediated remodeling of CL by examining the structural factors responsible for the superior acyl donor ability of dipalmitoleoyl (16:1) PC over dipalmitoyl (16:0) PC. To this end, we synthesized systematic derivatives of dipalmitoleoyl PC; for example, the location of the cis double bond was migrated from the Δ9-position toward either end of the acyl chains (the Δ5- or Δ13-position), the cis double bond in the sn-1 or sn-2 position or both, was changed to a trans form, and palmitoleoyl and palmitoyl groups were exchanged in the sn-1 and sn-2 positions, maintaining similar PC fluidities. Analyses of the tafazzin-mediated transacylation from these PCs to sn-2'-MLCL(18:1-18:1/18:1-OH) in the liposomal membrane revealed that tafazzin strictly discriminates the molecular configuration of the acyl chains of PCs, including their glycerol positions (sn-1 or sn-2); however, the effects of PC fluidity on the reaction may not be neglected. On the basis of the findings described herein, we discuss the relevance of the so-called thermodynamic remodeling hypothesis that presumes no acyl selectivity of tafazzin.
Product Description
Avanti
16:0-18:1 PC, 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine, powder
Avanti
16:0-18:1 PC, 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine, chloroform