- Simplifying microfluidic separation devices towards field-detection of blood parasites
Simplifying microfluidic separation devices towards field-detection of blood parasites
Analytical Methods : Advancing Methods and Applications (2016)
Holm SH, et al.