Merck
  • Abnormalities of prothrombin: a review of the pathophysiology, diagnosis, and treatment
Abnormalities of prothrombin: a review of the pathophysiology, diagnosis, and treatment
Haemophilia : the Official Journal of the World Federation of Hemophilia (2008)
Meeks, SL and Abshire, TC