Merck
  • Development of a Forster resonance energy transfer assay for monitoring bacterial collagenase triple-helical peptidase activity
Development of a Forster resonance energy transfer assay for monitoring bacterial collagenase triple-helical peptidase activity
Analytical biochemistry (2014)
Tokmina-Roszyk M, et al.