Merck
  • Protein kinase C activation causes neurite retraction via cyclin D 1 and p60-katanin increase in rat hippocampal neurons
Protein kinase C activation causes neurite retraction via cyclin D 1 and p60-katanin increase in rat hippocampal neurons
The European Journal of Neuroscience (2013)
Korulu S, et al.