Merck
  • Necrostatin-1 protects against glutamate-induced glutathione depletion and caspase-independent cell death in HT-22 cells
Necrostatin-1 protects against glutamate-induced glutathione depletion and caspase-independent cell death in HT-22 cells
Journal of Neurochemistry (2007)
Xu X, et al.