Merck
  • Carboxymethylcellulose binds to human corneal epithelial cells and is a modulator of corneal epithelial wound healing
Carboxymethylcellulose binds to human corneal epithelial cells and is a modulator of corneal epithelial wound healing
Investigative Ophthalmology & Visual Science (2007)
Garrett Q, et al.