Merck
  • Metabolic and histologic effects of sodium pyruvate treatment in the rat after cortical contusion injury
Metabolic and histologic effects of sodium pyruvate treatment in the rat after cortical contusion injury
Journal of Neurotrauma (2009)
Fukushima M, et al.