Merck
  • Preparation and evaluation of multifunctional envelope-type nano device loading siRNA
Preparation and evaluation of multifunctional envelope-type nano device loading siRNA
Artificial Cells, Nanomedicine, and Biotechnology (Print) (2016)
Yin H, et al.