Merck
  • Synthesis and characterization of TM-MCM-48 (TM= Mn, V, Cr) and their catalytic activity in the oxidation of styrene.
Synthesis and characterization of TM-MCM-48 (TM= Mn, V, Cr) and their catalytic activity in the oxidation of styrene.
J. Catal. (2005)
Gomez S, et al.