- Interaction of 4?-6-diamidino-2-phenylindole to nucleic acids, and its implication to their template activity in RNA-polymerase reaction of E. coli bacteria and of friend-virus infected mouse spleen
Interaction of 4?-6-diamidino-2-phenylindole to nucleic acids, and its implication to their template activity in RNA-polymerase reaction of E. coli bacteria and of friend-virus infected mouse spleen
Cancer letters (1978)
Mildner B, et al.