Merck
  • IL-4 together with IL-1β induces antitumor Th9 cell differentiation in the absence of TGF-β signaling.
IL-4 together with IL-1β induces antitumor Th9 cell differentiation in the absence of TGF-β signaling.
Nature communications (2019-03-28)
Gang Xue, Guangxu Jin, Jing Fang, Yong Lu
ABSTRACT
IL-9-producing CD4+ (Th9) cells are a subset of CD4+ T-helper cells that are endowed with powerful antitumor capacity. Both IL-4 and TGF-β have been reported to be indispensable for Th9 cell-priming and differentiation. Here we show, by contrast, that Th9 cell development can occur in the absence of TGF-β signaling. When TGF-β was replaced by IL-1β, the combination of IL-1β and IL-4 efficiently promoted IL-9-producing T cells (Th9IL-4+IL-1β). Th9IL-4+ IL-1β cells are phenotypically distinct T cells compared to classic Th9 cells (Th9IL-4+TGF-β) and other Th cells, and are enriched for IL-1 and NF-κB gene signatures. Inhibition of NF-κB but not TGF-β-signaling negates IL-9 production by Th9IL-4+IL-1β cells. Furthermore, when compared with classic Th9IL-4+TGF-β cells, Th9IL-4+IL-1β cells are less exhausted, exhibit cytotoxic T effector gene signature and tumor killing function, and exert a superior antitumor response in a mouse melanoma model. Our study thus describes an alternative pathway for Th9 cell differentiation and provides a potential avenue for antitumor therapies.
Sigma-Aldrich
TGF-β RI Kinase Inhibitor II, TGF-β RI Kinase Inhibitor II, CAS 446859-33-2, is a cell-permeable, potent, reversible, ATP-competitive inhibitor of TGF-β R1 kinase (IC50 = 23 nM and 4 nM for ALK5 binding & auto-phosphorylation).