Merck
  • Receptor tyrosine kinases (RTKs) consociate in regulatory clusters in Alzheimer's disease and type 2 diabetes.
Receptor tyrosine kinases (RTKs) consociate in regulatory clusters in Alzheimer's disease and type 2 diabetes.
Molecular and cellular biochemistry (2019-06-04)
Piyali Majumder, Kasturi Roy, Sangram Bagh, Debashis Mukhopadhyay
ABSTRACT
Alzheimer's disease (AD) and type 2 diabetes (T2D) share the common hallmark of insulin resistance. It is conjectured that receptor tyrosine kinases (RTKs) play definitive roles in the process. To decipher the signaling overlap behind this phenotypic resemblance, the activity status of RTKs is probed in post-mortem AD and T2D tissues and cell models. Activities of only about one-third changed in a similar fashion, whereas about half of them showed opposite outcomes when exposed to contrasting signals akin to AD and T2D. Interestingly, irrespective of disease type, RTKs with enhanced and compromised activities clustered distinctly, indicating separate levels of regulations. Similar regulatory mechanisms within an activity cluster could be inferred, which have potential to impact future therapeutic developments.