Merck
  • MiRNA-27a sensitizes breast cancer cells to treatment with Selective Estrogen Receptor Modulators.
MiRNA-27a sensitizes breast cancer cells to treatment with Selective Estrogen Receptor Modulators.
Breast (Edinburgh, Scotland) (2018-11-12)
Bojan Ljepoja, Jonathan García-Roman, Ann-Katrin Sommer, Ernst Wagner, Andreas Roidl
ABSTRACT
MicroRNA-27a (miR-27a) is a small non-coding RNA, shown to play a role in multiple cancers, including the regulation of ERα expression in breast cancer. Most ERα positive tumors are treated with Selective Estrogen Receptor Modulators (SERMs) and thus the role of miR-27a expression in response to SERM treatment is of interest. Tamoxifen resistant cells were generated by molecular evolution with six cycles of tamoxifen treatment. MCF7 and T47D luminal A breast cancer cell lines were either treated with miR-27a mimics, or ER-signaling was modulated ectopically. The changes were analyzed with RT-qPCR, western blotting and transcriptional activity ERE-reporter assays. Moreover, the response to SERM treatments (tamoxifen, endoxifen and toremifen) was investigated by cell viability and apoptosis measurements. An in silico analysis of survival data from the METABRIC study was performed in order to assess the prognostic value of miR-27a for response to SERM treatment. Tamoxifen-resistant cells showed decreased expression of ERα and miR-27a. The overexpression of miR-27a increased the levels of ERα, while modulation of ERα decreased miR-27a expression. High miR-27a expression increased the sensitivity of MCF7 and T47D cells to SERM treatments and re-sensitized the cells to tamoxifen. Patient survival of luminal A breast cancer patients that underwent endocrine therapies was better in groups with high miR-27a expression. MiR-27a sensitizes luminal A breast cancer cells to SERM treatments based on a positive feedback loop with ERα. An increased overall-survival of ER-positive breast cancer patients that underwent endocrine treatments and displayed high miR-27a levels was found.
Sigma-Aldrich
(E/Z)-Endoxifen Hydrochloride Hydrate, ≥98% (HPLC)
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Toremifene citrate salt, ≥98% (HPLC)
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture