Merck
  • Blood vessels guide Schwann cell migration in the adult demyelinated CNS through Eph/ephrin signaling.
Blood vessels guide Schwann cell migration in the adult demyelinated CNS through Eph/ephrin signaling.
Acta neuropathologica (2019-04-24)
Beatriz Garcia-Diaz, Corinne Bachelin, Fanny Coulpier, Gaspard Gerschenfeld, Cyrille Deboux, Violetta Zujovic, Patrick Charnay, Piotr Topilko, Anne Baron-Van Evercooren
ABSTRACT
Schwann cells (SC) enter the central nervous system (CNS) in pathophysiological conditions. However, how SC invade the CNS to remyelinate central axons remains undetermined. We studied SC migratory behavior ex vivo and in vivo after exogenous transplantation in the demyelinated spinal cord. The data highlight for the first time that SC migrate preferentially along blood vessels in perivascular extracellular matrix (ECM), avoiding CNS myelin. We demonstrate in vitro and in vivo that this migration route occurs by virtue of a dual mode of action of Eph/ephrin signaling. Indeed, EphrinB3, enriched in myelin, interacts with SC Eph receptors, to drive SC away from CNS myelin, and triggers their preferential adhesion to ECM components, such as fibronectin via integrinβ1 interactions. This complex interplay enhances SC migration along the blood vessel network and together with lesion-induced vascular remodeling facilitates their timely invasion of the lesion site. These novel findings elucidate the mechanism by which SC invade and contribute to spinal cord repair.
Sigma-Aldrich
Anti-EPHB6 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, clone 211F1.1, from mouse
Sigma-Aldrich
Monoclonal Anti-Fibronectin, Cellular antibody produced in mouse, clone FN-3E2, ascites fluid
Sigma-Aldrich
Anti-Neurofilament 200 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-EPHB1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-GLUT-1 Antibody, CT, from rabbit, purified by affinity chromatography