Merck
CD8+ T cells induce cachexia during chronic viral infection.
Nature immunology (2019-05-22)
Hatoon Baazim, Martina Schweiger, Michael Moschinger, Haifeng Xu, Thomas Scherer, Alexandra Popa, Suchira Gallage, Adnan Ali, Kseniya Khamina, Lindsay Kosack, Bojan Vilagos, Mark Smyth, Alexander Lercher, Joachim Friske, Doron Merkler, Alan Aderem, Thomas H Helbich, Mathias Heikenwälder, Philipp A Lang, Rudolf Zechner, Andreas Bergthaler
ABSTRACT
Cachexia represents a leading cause of morbidity and mortality in various cancers, chronic inflammation and infections. Understanding of the mechanisms that drive cachexia has remained limited, especially for infection-associated cachexia (IAC). In the present paper we describe a model of reversible cachexia in mice with chronic viral infection and identify an essential role for CD8+ T cells in IAC. Cytokines linked to cancer-associated cachexia did not contribute to IAC. Instead, virus-specific CD8+ T cells caused morphologic and molecular changes in the adipose tissue, which led to depletion of lipid stores. These changes occurred at a time point that preceded the peak of the CD8+ T cell response and required T cell-intrinsic type I interferon signaling and antigen-specific priming. Our results link systemic antiviral immune responses to adipose-tissue remodeling and reveal an underappreciated role of CD8+ T cells in IAC.
Sigma-Aldrich
FTY720, ≥98% (HPLC)