Merck
  • An alternative splicing switch in FLNB promotes the mesenchymal cell state in human breast cancer.
An alternative splicing switch in FLNB promotes the mesenchymal cell state in human breast cancer.
eLife (2018-07-31)
Ji Li, Peter S Choi, Christine L Chaffer, Katherine Labella, Justin H Hwang, Andrew O Giacomelli, Jong Wook Kim, Nina Ilic, John G Doench, Seav Huong Ly, Chao Dai, Kimberly Hagel, Andrew L Hong, Ole Gjoerup, Shom Goel, Jennifer Y Ge, David E Root, Jean J Zhao, Angela N Brooks, Robert A Weinberg, William C Hahn
ABSTRACT
Alternative splicing of mRNA precursors represents a key gene expression regulatory step and permits the generation of distinct protein products with diverse functions. In a genome-scale expression screen for inducers of the epithelial-to-mesenchymal transition (EMT), we found a striking enrichment of RNA-binding proteins. We validated that QKI and RBFOX1 were necessary and sufficient to induce an intermediate mesenchymal cell state and increased tumorigenicity. Using RNA-seq and eCLIP analysis, we found that QKI and RBFOX1 coordinately regulated the splicing and function of the actin-binding protein FLNB, which plays a causal role in the regulation of EMT. Specifically, the skipping of FLNB exon 30 induced EMT by releasing the FOXC1 transcription factor. Moreover, skipping of FLNB exon 30 is strongly associated with EMT gene signatures in basal-like breast cancer patient samples. These observations identify a specific dysregulation of splicing, which regulates tumor cell plasticity and is frequently observed in human cancer.
Sigma-Aldrich
Anti-A2BP1 (ab1) antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Filamin B Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.