Merck
  • A Novel Thermostable GH3 beta-Glucosidase from Talaromyce leycettanus with Broad Substrate Specificity and Significant Soybean Isoflavone Glycosides-Hydrolyzing Capability
A Novel Thermostable GH3 beta-Glucosidase from Talaromyce leycettanus with Broad Substrate Specificity and Significant Soybean Isoflavone Glycosides-Hydrolyzing Capability
BioMed Research International (2018)
Li X, et al.