ES
EN
Applications
Products
Services
Support
Home
Search Results
Determination of folic acid in tablets by microemulsion electrokinetic chromatography
Determination of folic acid in tablets by microemulsion electrokinetic chromatography
Journal of Chromatography A (2004)
Aurora-Prado SM, et al.