- HPLC-MS/MS method for simultaneous quantification of vildagliptin, metformin, and metformin-related compounds in tablets
HPLC-MS/MS method for simultaneous quantification of vildagliptin, metformin, and metformin-related compounds in tablets
International Journal of Pharmacy and Pharmaceutical Sciences (2014)
Uber CP, et al.