Merck
  • Formation of HETE-EAs and dihydroxy derivatives in mouse kidney tissue and analysis by high-performance liquid chromatography tandem mass spectrometry.
Formation of HETE-EAs and dihydroxy derivatives in mouse kidney tissue and analysis by high-performance liquid chromatography tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2019-08-23)
Sara K Dempsey, Ashley M Gesseck, Ashfaq Ahmad, Zdravka Daneva, Joseph K Ritter, Justin L Poklis
ABSTRACT
The kidneys play an important role in the long-term regulation of blood pressure by control of salt and water balance in the body through various systems including the endocannabinoid system. The endocannabinoid system consists of the two major cannabinoid receptor agonists, anandamide (AEA) and 2-arachidonylglycerol (2-AG), their hydrolyzing enzymes, fatty acid amide hydrolase (FAAH) and monoacylglycerol lipase (MAGL), and the cannabinoid receptors, CB1 and CB2. AEA can be converted into 12- and 15(S)-hydroperoxyeicosatetraenoic acid ethanolamides by 12-LOX and 15-LOX, respectively and can form epoxyeicosatrienoic acid- (EET-EAs) (5,6-, 8,9-, 11,12-, 14,15-) and hydroxyeicosatetraenoic acid- (HETE) ethanolamides. Furthermore, the EET-EAs produce a secondary metabolism by microsomal epoxide hydrolase to form the corresponding dihydroxyeicosatetraenoic acid-EAs (DiHETE-EA). Reference material was not available for DiHETE-EA. These metabolites were synthesized by incubation of the corresponding EET-EAs with mouse liver cytosol containing epoxide hydrolases. Presented is a solid phase extraction and high-performance liquid chromatography tandem mass spectrometry (HPLC-MS/MS) for the extraction and quantitation of AEA, 2-AG, their metabolites, oleoylethanolamide (OEA), and palmitoylethanolamide (PEA), and the in vivo formation of the DiHETE-EAs in kidney after a single intravenous bolus administration of 20 mg/kg of anandamide in C57BL/6 J and FAAH KO mice.
Sigma-Aldrich
Arachidonylethanolamide, ≥97.0% (TLC), oil
Sigma-Aldrich
N-Oleoylethanolamine, ~98% (TLC)