Merck
  • TROSY in NMR studies of the structure and function of large biological macromolecules.
TROSY in NMR studies of the structure and function of large biological macromolecules.
Current opinion in structural biology (2003-10-22)
César Fernández, Gerhard Wider
ABSTRACT
Transverse relaxation-optimized spectroscopy (TROSY), in combination with various isotope-labeling techniques, has opened avenues to study biomolecules with molecular masses of up to 1000000Da by solution NMR. Important recent applications of TROSY include the structure determination of membrane proteins in detergent micelles, structural and functional studies of large proteins in both monomeric form and macromolecular complexes, and investigations of intermolecular interactions in large complexes. TROSY improves the measurement of residual dipolar couplings and the detection of scalar couplings across hydrogen bonds - techniques that promise to further enhance the determination of solution structures of large proteins and oligonucleotides.
Sigma-Aldrich
SLAM-Aβ-13C3 Methyl Labeling Kit
Sigma-Aldrich
HLAM-AβIδ1MεLVproSTγ-13CH3 Methyl Labeling Kit