- An integrated workflow for crosslinking mass spectrometry.
An integrated workflow for crosslinking mass spectrometry.
Molecular systems biology (2019-09-27)
Marta L Mendes, Lutz Fischer, Zhuo A Chen, Marta Barbon, Francis J O'Reilly, Sven H Giese, Michael Bohlke-Schneider, Adam Belsom, Therese Dau, Colin W Combe, Martin Graham, Markus R Eisele, Wolfgang Baumeister, Christian Speck, Juri Rappsilber
PMID31556486
ABSTRACT
We present a concise workflow to enhance the mass spectrometric detection of crosslinked peptides by introducing sequential digestion and the crosslink identification software xiSEARCH. Sequential digestion enhances peptide detection by selective shortening of long tryptic peptides. We demonstrate our simple 12-fraction protocol for crosslinked multi-protein complexes and cell lysates, quantitative analysis, and high-density crosslinking, without requiring specific crosslinker features. This overall approach reveals dynamic protein-protein interaction sites, which are accessible, have fundamental functional relevance and are therefore ideally suited for the development of small molecule inhibitors.
Sigma-Aldrich
Albumin from human serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Myoglobin from equine heart, ≥90% (SDS-PAGE), essentially salt-free, lyophilized powder