Merck
  • RNF41 (Nrdp1) controls type 1 cytokine receptor degradation and ectodomain shedding.
RNF41 (Nrdp1) controls type 1 cytokine receptor degradation and ectodomain shedding.
Journal of cell science (2011-03-08)
Joris Wauman, Leentje De Ceuninck, Nele Vanderroost, Sam Lievens, Jan Tavernier
ABSTRACT
Cytokines, such as interferons, erythropoietin, leptin and most interleukins, signal through type 1 cytokine receptors and activate the canonical JAK-STAT pathway. Aberrant cytokine signalling underlies numerous pathologies and adequate, temporary receptor activation is therefore under tight control. Negative-feedback mechanisms are very well studied, but cellular sensitivity also depends on the number of receptors exposed at the cell surface. This is determined by the equilibrium between receptor synthesis and transport to the plasma membrane, internalisation and recycling, degradation and ectodomain shedding, but the molecular basis of how cells establish steady state receptor levels is poorly understood. Here, we report that ring finger protein 41 (RNF41, also known as E3 ubiquitin-protein ligase Nrdp1) interacts with JAK2-associated cytokine receptor complexes and modulates their cell surface exposure and signalling. Moreover, ectopic expression of RNF41 affected turnover of leptin, leukaemia inhibitory factor and interleukin-6 receptor in a dual way: it blocked intracellular cathepsin-L-dependent receptor cleavage and concomitantly enhanced receptor shedding by metalloproteases of the ADAM family. Receptor degradation and shedding are thus interconnected phenomena with a single protein, RNF41, determining the balance.
Sigma-Aldrich
CA-074 Me, CA-074 Me, CAS 147859-80-1, is a cell-permeable analog of CA-074 that acts as an irreversible inhibitor of intracellular cathepsin B.
Sigma-Aldrich
Actin from rabbit muscle, ≥85% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP), meets EP testing specifications
Sigma-Aldrich
Ponceau S, Dye content 75 %
Sigma-Aldrich
Ponceau S, BioReagent, suitable for electrophoresis
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder