Merck
  • Comparison of RNA isolation procedures for analysis of adult murine brain and spinal cord astrocytes.
Comparison of RNA isolation procedures for analysis of adult murine brain and spinal cord astrocytes.
Journal of neuroscience methods (2019-12-11)
Kamil Sebastian Rosiewicz, Tadhg Crowley, Gesine Saher, Janis Kerkering, Marlen Alisch, Volker Siffrin
ABSTRACT
Molecular analyses of cell populations and single cells have been instrumental in the advancement of our understanding of the physiology and pathologic processes of the nervous system. However, the limitation of these methods is the dependence on a gentle, efficient and specific enrichment procedure for the target cell population. In particular, this has been challenging for tightly interconnected cells, for example central nervous system (CNS) endogenous cells such as astrocytes. Here we adopted one of the most common methods of cell extraction, namely, enzymatic tissue digestion followed by fluorescence-activated cell sorting (FACS) of individual cells. We evaluated different enzymatic/mechanical tissue dissociation procedures and analyzed different astrocyte lineage transgenic models. Furthermore, we compared the cell extraction efficiency from spinal cord vs. brain. Enzymatic digestion of CNS tissue of Glast-CreERT2x tdTomatofl/fl or Aldh1l1-CreERT2x tdTomatofl/fl followed by FACS resulted in highly purified astrocytes. Automated tissue digestion strongly improved the isolated cell numbers. Aldh1l1-CreERT2 identified more astrocytes than Glast-CreERT2; isolation from brain yields higher numbers than from spinal cord. We compared the efficiency and purity of the enzymatic dissociation/FACS approach with a more modern procedure consisting of tissue homogenization followed by translating ribosome affinity purification (TRAP). We found that both methods result in highly enriched astrocytic RNA. However, only TRAP isolation resulted in reliably detectable RNA concentrations from spinal cord tissue on a single animal level. Depending on the aim of the study both methods have advantages and disadvantages but both are acceptable for astrocytic RNA analysis.
Product Description
Avanti
07:0 PC (DHPC), 1,2-diheptanoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
07:0 PC (DHPC), Avanti Research - A Croda Brand
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)