Merck
  • A toolbox of immunoprecipitation-grade monoclonal antibodies to human transcription factors.
A toolbox of immunoprecipitation-grade monoclonal antibodies to human transcription factors.
Nature methods (2018-04-12)
Anand Venkataraman, Kun Yang, Jose Irizarry, Mark Mackiewicz, Paolo Mita, Zheng Kuang, Lin Xue, Devlina Ghosh, Shuang Liu, Pedro Ramos, Shaohui Hu, Diane Bayron Kain, Sarah Keegan, Richard Saul, Simona Colantonio, Hongyan Zhang, Florencia Pauli Behn, Guang Song, Edisa Albino, Lillyann Asencio, Leonardo Ramos, Luvir Lugo, Gloriner Morell, Javier Rivera, Kimberly Ruiz, Ruth Almodovar, Luis Nazario, Keven Murphy, Ivan Vargas, Zully Ann Rivera-Pacheco, Christian Rosa, Moises Vargas, Jessica McDade, Brian S Clark, Sooyeon Yoo, Seva G Khambadkone, Jimmy de Melo, Milanka Stevanovic, Lizhi Jiang, Yana Li, Wendy Y Yap, Brittany Jones, Atul Tandon, Elliot Campbell, Gaetano T Montelione, Stephen Anderson, Richard M Myers, Jef D Boeke, David Fenyö, Gordon Whiteley, Joel S Bader, Ignacio Pino, Daniel J Eichinger, Heng Zhu, Seth Blackshaw
ABSTRACT
A key component of efforts to address the reproducibility crisis in biomedical research is the development of rigorously validated and renewable protein-affinity reagents. As part of the US National Institutes of Health (NIH) Protein Capture Reagents Program (PCRP), we have generated a collection of 1,406 highly validated immunoprecipitation- and/or immunoblotting-grade mouse monoclonal antibodies (mAbs) to 737 human transcription factors, using an integrated production and validation pipeline. We used HuProt human protein microarrays as a primary validation tool to identify mAbs with high specificity for their cognate targets. We further validated PCRP mAbs by means of multiple experimental applications, including immunoprecipitation, immunoblotting, chromatin immunoprecipitation followed by sequencing (ChIP-seq), and immunohistochemistry. We also conducted a meta-analysis that identified critical variables that contribute to the generation of high-quality mAbs. All validation data, protocols, and links to PCRP mAb suppliers are available at http://proteincapture.org.
Product Description
Millipore
Protein G–Agarose, lyophilized powder, Contains lactose stabilizers that must be removed prior to use.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)