  • Search Results
  • Pulmonary surfactant protein SP-B nanorings induce the multilamellar organization of surfactant complexes.
Pulmonary surfactant protein SP-B nanorings induce the multilamellar organization of surfactant complexes.
Biochimica et biophysica acta. Biomembranes (2020-02-19)
Marta Martinez-Calle, Manuel Prieto, Bárbara Olmeda, Aleksander Fedorov, Luís M S Loura, Jesús Pérez-Gil
ABSTRACT
Surfactant protein SP-B is absolutely required for the generation of functional pulmonary surfactant, a unique network of multilayered membranes, which stabilizes the respiratory air-liquid interface. It has been proposed that SP-B assembles into hydrophobic rings and tubes that facilitate the rapid transfer of phospholipids from membrane stores into the interface and the formation of multilayered films, ensuring the stability of the alveoli against physical forces leading to their collapse. To elucidate the molecular organization of SP-B-promoted multilamellar membrane structures, time-resolved Förster Resonance Energy Transfer (FRET) experiments between BODIPY-PC or BODIPY-derivatized SP-B (BODIPY/SP-B), as donor probes, and octadecylrhodamine B, as acceptor probe, were performed in liposomes containing SP-B or BODIPY/SP-B. Our results show that both SP-B and fluorescently labeled SP-B oligomers mediate the connection of adjacent bilayers. Furthermore, by applying rational models to the FRET data, we have been able to provide quantitative details of the structure of SP-B-induced multilayered membrane arrays at the nanometer scale, defining interactions between SP-B rings as key elements for connecting surfactant membranes. The data sustain the structural model and the mechanism of action of SP-B assemblies to sustain the crucial surfactant function.
Product Description
Avanti
16:0-18:1 PG, 1-palmitoyl-2-oleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), powder
Avanti
16:0-18:1 PG, 1-palmitoyl-2-oleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), chloroform
Avanti
16:0-18:1 PC, 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine, powder
Avanti
16:0-18:1 PC, 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine, chloroform