- Manganese (II) complexes with a bulky anthracene-based dicarboxylic ligand: Syntheses, crystal structures, and magnetic properties
Manganese (II) complexes with a bulky anthracene-based dicarboxylic ligand: Syntheses, crystal structures, and magnetic properties
Australian Journal of Chemistry (2008)
Liu C, et al.