Merck
Development and Characterization of a Wee1 Kinase Degrader.
Cell chemical biology (2019-11-19)
Zhengnian Li, Benika J Pinch, Calla M Olson, Katherine A Donovan, Radosław P Nowak, Caitlin E Mills, David A Scott, Zainab M Doctor, Nicholas A Eleuteri, Mirra Chung, Peter K Sorger, Eric S Fischer, Nathanael S Gray
ABSTRACT
The G1/S cell cycle checkpoint is frequently dysregulated in cancer, leaving cancer cells reliant on a functional G2/M checkpoint to prevent excessive DNA damage. Wee1 regulates the G2/M checkpoint by phosphorylating CDK1 at Tyr15 to prevent mitotic entry. Previous drug development efforts targeting Wee1 resulted in the clinical-grade inhibitor, AZD1775. However, AZD1775 is burdened by dose-limiting adverse events, and has off-target PLK1 activity. In an attempt to overcome these limitations, we developed Wee1 degraders by conjugating AZD1775 to the cereblon (CRBN)-binding ligand, pomalidomide. The resulting lead compound, ZNL-02-096, degrades Wee1 while sparing PLK1, induces G2/M accumulation at 10-fold lower doses than AZD1775, and synergizes with Olaparib in ovarian cancer cells. We demonstrate that ZNL-02-096 has CRBN-dependent pharmacology that is distinct from AZD1775, which justifies further evaluation of selective Wee1 degraders.
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
3-Chloroperbenzoic acid, ≤77%
Sigma-Aldrich
Lenalidomide, ≥95%
Sigma-Aldrich
Pomalidomide, ≥98% (HPLC)