Merck
  • Direct visualization of degradation microcompartments at the ER membrane.
Direct visualization of degradation microcompartments at the ER membrane.
Proceedings of the National Academy of Sciences of the United States of America (2019-12-29)
Sahradha Albert, Wojciech Wietrzynski, Chia-Wei Lee, Miroslava Schaffer, Florian Beck, Jan M Schuller, Patrice A Salomé, Jürgen M Plitzko, Wolfgang Baumeister, Benjamin D Engel
ABSTRACT
To promote the biochemical reactions of life, cells can compartmentalize molecular interaction partners together within separated non-membrane-bound regions. It is unknown whether this strategy is used to facilitate protein degradation at specific locations within the cell. Leveraging in situ cryo-electron tomography to image the native molecular landscape of the unicellular alga Chlamydomonas reinhardtii, we discovered that the cytosolic protein degradation machinery is concentrated within ∼200-nm foci that contact specialized patches of endoplasmic reticulum (ER) membrane away from the ER-Golgi interface. These non-membrane-bound microcompartments exclude ribosomes and consist of a core of densely clustered 26S proteasomes surrounded by a loose cloud of Cdc48. Active proteasomes in the microcompartments directly engage with putative substrate at the ER membrane, a function canonically assigned to Cdc48. Live-cell fluorescence microscopy revealed that the proteasome clusters are dynamic, with frequent assembly and fusion events. We propose that the microcompartments perform ER-associated degradation, colocalizing the degradation machinery at specific ER hot spots to enable efficient protein quality control.
Sigma-Aldrich
NMS-873, ≥98% (HPLC)
Sigma-Aldrich
MG-132, ≥95% by HPLC, Potent, reversible, and cell-permeable proteasome inhibitor (Ki = 4 nM).
Sigma-Aldrich
Tunicamycin from Streptomyces sp.