Merck
  • The m6A methyltransferase METTL3 cooperates with demethylase ALKBH5 to regulate osteogenic differentiation through NF-κB signaling.
The m6A methyltransferase METTL3 cooperates with demethylase ALKBH5 to regulate osteogenic differentiation through NF-κB signaling.
Molecular and cellular biochemistry (2019-10-24)
Jinjin Yu, Lujun Shen, Yanli Liu, Hong Ming, Xinxing Zhu, Maoping Chu, Juntang Lin
ABSTRACT
As a m6A methylation modifier, METTL3 is functionally involved in various biological processes. Nevertheless, the role of METTL3 in osteogenesis is not determined up to date. In the current study, METTL3 is identified as a crucial regulator in the progression of osteogenic differentiation. Loss of METTL3 significantly augments calcium deposition and enhances alkaline phosphatase activity of mesenchymal stem cells, uncovering an inhibitory role of METTL3 in osteogenesis. More importantly, the underlying molecular basis by which METTL3 regulates osteogenesis is illustrated. We find that METTL3 positively regulates expression of MYD88, a critical upstream regulator of NF-κB signaling, by facilitating m6A methylation modification to MYD88-RNA, subsequently inducing the activation of NF-κB which is widely regarded as a repressor of osteogenesis and therefore suppressing osteogenic progression. Moreover, the METTL3-mediated m6A methylation is found to be dynamically reversed by the demethylase ALKBH5. In summary, this study highlights the functional importance of METTL3 in osteogenic differentiation and METTL3 may serve as a promising molecular target in regenerative medicine, as well as in the field of bone tissue engineering.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Alizarin Red S, certified by the BSC
Sigma-Aldrich
BCIP®/NBT Liquid Substrate System, ready to use solution
Sigma-Aldrich
Bay 11-7082, ≥98% (HPLC), powder