Merck
  • Reactive Extrusion of Nonmigratory Antioxidant Poly(lactic acid) Packaging.
Reactive Extrusion of Nonmigratory Antioxidant Poly(lactic acid) Packaging.
Journal of agricultural and food chemistry (2020-01-28)
Joshua E Herskovitz, Julie M Goddard
ABSTRACT
Reactive extrusion of bio-derived active packaging offers a new approach to address converging concerns over environmental contamination and food waste. Herein, metal-chelating nitrilotriacetic acid (NTA) ligands were grafted onto poly(lactic acid) (PLA) by reactive extrusion to produce metal-chelating PLA (PLA-g-NTA). Radical grafting was confirmed by attenuated total reflectance Fourier transform infrared (ATR-FTIR) spectroscopy with the introduction of secondary alkyl stretches (2919 and 2860 cm-1) and by X-ray photoelectron spectroscopy (XPS) with an increase in the atomic percentage of nitrogen. Compared to films prepared from native, granular PLA (gPLA), PLA-g-NTA films had lower contact angles and hysteresis values (86.35° ± 2.49 and 31.89° ± 2.27 to 79.91° ± 1.58 and 21.79° ± 1.72, respectively), supporting the surface orientation of the NTA ligands. The PLA-g-NTA films exhibited a significant antioxidant character with a radical scavenging capacity of 0.675 ± 0.026 nmol Trolox(eq)/cm2 and an iron chelation capacity of 54.09 ± 9.36 nmol/cm2. PLA-g-NTA films delayed ascorbic acid degradation, retaining ∼45% ascorbic acid over the 9-day study compared to <20% for control PLA. This research makes significant advances in translating active packaging technologies to bio-derived materials using scalable, commercially translatable synthesis methods.
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%
Sigma-Aldrich
Nitrilotriacetic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Dicumyl peroxide, 98%