- Single, binary and multi-component adsorption of copper and cadmium from aqueous solutions on Kraft lignin?a biosorbent
Single, binary and multi-component adsorption of copper and cadmium from aqueous solutions on Kraft lignin?a biosorbent
Journal of Colloid and Interface Science (2006)
Mohan D, et al.