Merck
  • Ubiquitination of DNA Damage-Stalled RNAPII Promotes Transcription-Coupled Repair.
Ubiquitination of DNA Damage-Stalled RNAPII Promotes Transcription-Coupled Repair.
Cell (2020-03-07)
Yuka Nakazawa, Yuichiro Hara, Yasuyoshi Oka, Okiru Komine, Diana van den Heuvel, Chaowan Guo, Yasukazu Daigaku, Mayu Isono, Yuxi He, Mayuko Shimada, Kana Kato, Nan Jia, Satoru Hashimoto, Yuko Kotani, Yuka Miyoshi, Miyako Tanaka, Akira Sobue, Norisato Mitsutake, Takayoshi Suganami, Akio Masuda, Kinji Ohno, Shinichiro Nakada, Tomoji Mashimo, Koji Yamanaka, Martijn S Luijsterburg, Tomoo Ogi
ABSTRACT
Transcription-coupled nucleotide excision repair (TC-NER) is initiated by the stalling of elongating RNA polymerase II (RNAPIIo) at DNA lesions. The ubiquitination of RNAPIIo in response to DNA damage is an evolutionarily conserved event, but its function in mammals is unknown. Here, we identified a single DNA damage-induced ubiquitination site in RNAPII at RPB1-K1268, which regulates transcription recovery and DNA damage resistance. Mechanistically, RPB1-K1268 ubiquitination stimulates the association of the core-TFIIH complex with stalled RNAPIIo through a transfer mechanism that also involves UVSSA-K414 ubiquitination. We developed a strand-specific ChIP-seq method, which revealed RPB1-K1268 ubiquitination is important for repair and the resolution of transcriptional bottlenecks at DNA lesions. Finally, RPB1-K1268R knockin mice displayed a short life-span, premature aging, and neurodegeneration. Our results reveal RNAPII ubiquitination provides a two-tier protection mechanism by activating TC-NER and, in parallel, the processing of DNA damage-stalled RNAPIIo, which together prevent prolonged transcription arrest and protect against neurodegeneration.
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-5) Antibody, clone 3E8, clone 3E8, from rat
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho CTD Ser-2) Antibody, clone 3E10, clone 3E10, from rat
Sigma-Aldrich
5-Ethynyl uridine, ≥95%
Sigma-Aldrich
Protein A Agarose/Salmon Sperm DNA, 2.5 mL, for use in chromatin immunoprecipitations (ChIP assays)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
α-Amanitin, from Amanita phalloides, ≥85% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Di(N-succinimidyl) glutarate, ≥97.0% (CHN)
Sigma-Aldrich
Mordant Orange 1, Dye content 70 %
Sigma-Aldrich
Toluidine Blue, suitable for microscopy, 8.74% (ZN (THEORY))
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Millipore
Benzonase® Nuclease, Purity > 99%, Effective viscosity reduction and removal of nucleic acids from protein solutions