Merck
  • The Lifeact-EGFP mouse is a translationally controlled fluorescent reporter of T cell activation.
The Lifeact-EGFP mouse is a translationally controlled fluorescent reporter of T cell activation.
Journal of cell science (2020-02-12)
Jorge Luis Galeano Niño, Szun S Tay, Jacqueline L E Tearle, Jianling Xie, Matt A Govendir, Daryan Kempe, Jessica Mazalo, Alexander P Drew, Feyza Colakoglu, Sarah K Kummerfeld, Christopher G Proud, Maté Biro
ABSTRACT
It has become increasingly evident that T cell functions are subject to translational control in addition to transcriptional regulation. Here, by using live imaging of CD8+ T cells isolated from the Lifeact-EGFP mouse, we show that T cells exhibit a gain in fluorescence intensity following engagement of cognate tumour target cells. The GFP signal increase is governed by Erk1/2-dependent distal T cell receptor (TCR) signalling and its magnitude correlates with IFN-γ and TNF-α production, which are hallmarks of T cell activation. Enhanced fluorescence was due to increased translation of Lifeact-EGFP protein, without an associated increase in its mRNA. Activation-induced gains in fluorescence were also observed in naïve and CD4+ T cells from the Lifeact-EGFP reporter, and were readily detected by both flow cytometry and live cell microscopy. This unique, translationally controlled reporter of effector T cell activation simultaneously enables tracking of cell morphology, F-actin dynamics and activation state in individual migrating T cells. It is a valuable addition to the limited number of reporters of T cell dynamics and activation, and opens the door to studies of translational activity and heterogeneities in functional T cell responses in situ.
Sigma-Aldrich
7-Cyclopentyl-5-(4-phenoxyphenyl)-7H-pyrrolo[2,3‑d]pyrimidin-4-ylamine, ≥98% (HPLC)
Pricing and availability is not currently available.