Merck
  • Impact of wastewater effluent pollution on stream functioning: A whole-ecosystem manipulation experiment.
Impact of wastewater effluent pollution on stream functioning: A whole-ecosystem manipulation experiment.
Environmental pollution (Barking, Essex : 1987) (2019-12-16)
Olatz Pereda, Libe Solagaistua, Miren Atristain, Ioar de Guzmán, Aitor Larrañaga, Daniel von Schiller, Arturo Elosegi
ABSTRACT
The ecological effects of wastewater treatment plant (WWTP) effluents on stream ecosystems cause growing concern. However, it is difficult to assess these effects as most streams receiving WWTP effluents are also affected by other stressors. We performed a whole-ecosystem manipulation experiment following a BACI design (Before-After/Control-Impact) in order to exclude the influence of other potentially confounding factors. We diverted part of the effluent of a large tertiary urban WWTP into a small, unpolluted stream, and studied its effects on ecosystem structure and functioning over two years (i.e., one year before and one year after the effluent diversion). Although highly diluted (final concentration in the receiving stream averaged 3%), the effluent promoted biofilm chlorophyll-a and biomass (2.3 and 2.1 times, respectively), exo-enzymatic activities (phosphatase 2.2 and glucosidase 4.2 times) and invertebrate-mediated organic matter decomposition (1.4 times), but reduced phosphorus uptake capacity of the epilithic biofilm down to 0.5 of the initial values. Biofilm metabolism, reach-scale nutrient uptake and microbially-mediated organic matter decomposition were not affected. Our results indicate that even well treated and highly diluted WWTP effluents can also affect the structure of the biofilm community and stream ecosystem functioning.
Sigma-Aldrich
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)