Merck
  • An immunochromatographic test system for the determination of lincomycin in foodstuffs of animal origin.
An immunochromatographic test system for the determination of lincomycin in foodstuffs of animal origin.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2020-02-23)
Olga D Hendrickson, Elena A Zvereva, Demid S Popravko, Anatoly V Zherdev, Chuanlai Xu, Boris B Dzantiev
ABSTRACT
The aim of this study was to develop a rapid and sensitive immunochromatographic test system for the detection of lincomycin (LIN), which belongs to the lincosamide group of antibiotics and contaminates food products of animal origin. Two formats of immunochromatographic analysis (ICA) based on different approaches of introducing gold nanoparticles (GNPs) as a label were compared. It was demonstrated that an indirect ICA method where GNPs were conjugated with anti-species antibodies allowed the achievement of both instrumental and visual detection limits of LIN almost two orders of magnitude lower than those achieved in the standard direct ICA format. In the optimized conditions, the developed indirect ICA allowed for the detection of LIN within 15 min, with instrumental and visual detection limits of 8 pg/mL and 0.8 ng/mL. The assay showed 40% cross-reactivity to clindamycin (CLIN) as a structural analogue of LIN, with no interaction with antibiotics from other classes. The developed ICA was applied for LIN detection in a panel of food products. No treatment of cow milk was necessary before the analysis. For chicken eggs and honey, a simple procedure of preliminary sample preparation was developed, which fully prevented a matrix influence on the assay results. It was demonstrated that ICA could detect LIN in food products while preserving the same analytical characteristics as in the buffer. The analytical recoveries of LIN in foodstuffs were 93.8-125% with coefficients of variations of 5.3-14.0%.
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine dihydrochloride, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Triton X-100, laboratory grade