Merck
  • Characterization of Cep85--a new antagonist of Nek2A that is involved in the regulation of centrosome disjunction
Characterization of Cep85--a new antagonist of Nek2A that is involved in the regulation of centrosome disjunction
Journal of Cell Science (2015)
Chen C, et al.