Merck
  • Targeting mir128-3p alleviates myocardial insulin resistance and prevents ischemia-induced heart failure.
Targeting mir128-3p alleviates myocardial insulin resistance and prevents ischemia-induced heart failure.
eLife (2020-04-01)
Andrea Ruiz-Velasco, Min Zi, Susanne S Hille, Tayyiba Azam, Namrita Kaur, Juwei Jiang, Binh Nguyen, Karolina Sekeres, Pablo Binder, Lucy Collins, Fay Pu, Han Xiao, Kaomei Guan, Norbert Frey, Elizabeth J Cartwright, Oliver J Müller, Xin Wang, Wei Liu
ABSTRACT
Myocardial insulin resistance contributes to heart failure in response to pathological stresses, therefore, a therapeutic strategy to maintain cardiac insulin pathways requires further investigation. We demonstrated that insulin receptor substrate 1 (IRS1) was reduced in failing mouse hearts post-myocardial infarction (MI) and failing human hearts. The mice manifesting severe cardiac dysfunction post-MI displayed elevated mir128-3p in the myocardium. Ischemia-upregulated mir128-3p promoted Irs1 degradation. Using rat cardiomyocytes and human-induced pluripotent stem cell-derived cardiomyocytes, we elucidated that mitogen-activated protein kinase 7 (MAPK7, also known as ERK5)-mediated CCAAT/enhancer-binding protein beta (CEBPβ) transcriptionally represses mir128-3p under hypoxia. Therapeutically, functional studies demonstrated gene therapy-delivered cardiac-specific MAPK7 restoration or overexpression of CEBPβ impeded cardiac injury after MI, at least partly due to normalization of mir128-3p. Furthermore, inhibition of mir128-3p preserved Irs1 and ameliorated cardiac dysfunction post-MI. In conclusion, we reveal that targeting mir128-3p mitigates myocardial insulin resistance, thereafter slowing down the progression of heart failure post-ischemia.
Sigma-Aldrich
Periodic Acid-Schiff (PAS) Staining System
Sigma-Aldrich
Methyltrioctylammonium hydrogen sulfate, ≥95.0% (T)
Sigma-Aldrich
Nuclear Fast Red Solution
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Sigma-Aldrich
Lactate Dehydrogenase Activity Assay Kit, sufficient for 500 colorimetric tests
Sigma-Aldrich
Wnt Antagonist II, IWP-2, The Wnt Antagonist II, IWP-2, also referenced under CAS 686770-61-6, controls the biological activity of Wnt. This small molecule/inhibitor is primarily used for Cancer applications.
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) Bioluminescent Assay Kit, for ATP quantitation
Sigma-Aldrich
Bouin′s solution, histological fixative
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid
Sigma-Aldrich
Anti-phospho-MEF2C (pSer396) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-IRS1 (Tyr608) mouse/ (Tyr612) human Antibody, Upstate®, from rabbit