Merck
  • Suppression of a phospholipase D gene, OsPLDbeta1, activates defense responses and increases disease resistance in rice.
Suppression of a phospholipase D gene, OsPLDbeta1, activates defense responses and increases disease resistance in rice.
Plant physiology (2009-03-17)
Takeshi Yamaguchi, Masaharu Kuroda, Hiromoto Yamakawa, Taketo Ashizawa, Kazuyuki Hirayae, Leona Kurimoto, Tomonori Shinya, Naoto Shibuya
ABSTRACT
Phospholipase D (PLD) plays an important role in plants, including responses to abiotic as well as biotic stresses. A survey of the rice (Oryza sativa) genome database indicated the presence of 17 PLD genes in the genome, among which OsPLDalpha1, OsPLDalpha5, and OsPLDbeta1 were highly expressed in most tissues studied. To examine the physiological function of PLD in rice, we made knockdown plants for each PLD isoform by introducing gene-specific RNA interference constructs. One of them, OsPLDbeta1-knockdown plants, showed the accumulation of reactive oxygen species in the absence of pathogen infection. Reverse transcription-polymerase chain reaction and DNA microarray analyses revealed that the knockdown of OsPLDbeta1 resulted in the up-/down-regulation of more than 1,400 genes, including the induction of defense-related genes such as pathogenesis-related protein genes and WRKY/ERF family transcription factor genes. Hypersensitive response-like cell death and phytoalexin production were also observed at a later phase of growth in the OsPLDbeta1-knockdown plants. These results indicated that the OsPLDbeta1-knockdown plants spontaneously activated the defense responses in the absence of pathogen infection. Furthermore, the OsPLDbeta1-knockdown plants exhibited increased resistance to the infection of major pathogens of rice, Pyricularia grisea and Xanthomonas oryzae pv oryzae. These results suggested that OsPLDbeta1 functions as a negative regulator of defense responses and disease resistance in rice.
Product Description
Avanti
14:0-12:0 NBD PC, Avanti Research - A Croda Brand 810123P, powder
Pricing and availability is not currently available.