Merck
B7-H4 is a potential prognostic biomarker of prostate cancer.
Experimental and molecular pathology (2020-02-24)
Haoyue Li, Lihua Piao, Sicen Liu, Yan Cui, Yanhua Xuan
ABSTRACT
B7-H4 is a member of B7 family which regulates immune responses by delivering costimulatory signals. However, it negatively regulates T cell-mediated immunity and may play an important role in tumor immune evasion. Although several studies have been reported that expression of B7-H4 is elevated in the several types of human cancer with a poor clinical outcome, its clinical significance in the prostate cancer (PCa) has not been well studied. In this study, we investigated the clinical significance of B7-H4 in human PCa and determined if B7-H4 expression is associated with the cancer cell stemness in PCa. Our studies show that expression of B7-H4 is correlated with the pathologic tumor (pT) stage and the clinical stage of PCa. The Kaplan-Meier survival analysis revealed that PCa patients with high expression of B7-H4 exhibits a shorter overall survival (OS) rate. Univariate and multivariate Cox regression analysis indicated that B7-H4 is an independent poor prognostic factor of PCa. In addition, the expression of B7-H4 is correlated with the cancer cell stemness associated genes expression in PCa. Further, our studies show that B7-H4 regulates cancer cell stemness associated genes expression and effects on the cell cycle and PI3K/Akt signaling related genes expression in PCa. These results indicate that B7-H4 expression is associated with cancer cell stemness, and B7-H4 is a potential prognostic biomarker and a therapeutic target of PCa.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
MISSION® esiRNA, targeting human VTCN1