Merck
  • Proteoglycan IMPG2 Shapes the Interphotoreceptor Matrix and Modulates Vision.
Proteoglycan IMPG2 Shapes the Interphotoreceptor Matrix and Modulates Vision.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2020-04-09)
Ezequiel M Salido, Visvanathan Ramamurthy
ABSTRACT
Photoreceptor neurons are surrounded by an extracellular matrix, called the interphotoreceptor matrix (IPM). Activities crucial to vision occur within the IPM, including trafficking of nutrients and metabolites, retinal attachment, and interactions needed for normal outer segment phagocytosis. The IPM includes the following two unique proteoglycans: IPM proteoglycan 1 (IMPG1) and IMPG2. Patients with mutations in IMPG1/IMPG2 develop visual deficits with subretinal material accumulation, highlighting the critical role of the IPM in vision. To determine the role of these proteoglycans in retinal physiology and the pathologic mechanisms that lead to vision loss, we generated mouse models lacking IMPG1/IMPG2. In normal retina, IMPG1 and IMPG2 occupy distinct IPM compartments, represent the main source of chondroitin sulfate and are fundamental for the constitution of the cone-specific glycocalyx stained by the PNA (peanut agglutinin) lectin marker. No evident morphologic or functional deficits were found in mice lacking IMPG1. In the absence of IMPG2, IMPG1 abnormally accumulated at the subretinal space need, likely leading to the formation of subretinal lesions and reduced visual function. Interestingly, mice lacking both IMPG1 and IMPG2, regardless of sex, showed normal retinal structure and function, demonstrating that the aberrant IMPG1 distribution is the main cause of the visual alterations observed in the absence of IMPG2. In conclusion, our results show the dependence of secreted proteoglycans such as IMPG1 on the extracellular environment to properly integrate into the matrix, demonstrate the role of IMPG2 in shaping the IPM, and shed light on the potential mechanisms leading to the development of subretinal lesions and vision loss.SIGNIFICANCE STATEMENT The photoreceptors are specialized neurons that drive phototransduction in the mammalian retina. These cells are organized and surrounded by an extracellular matrix, the interphotoreceptor matrix (IPM). Mutations in IPM proteoglycans are associated with blindness in humans. Our studies show that two specific proteoglycans of the IPM, IPM proteoglycan 1 (IMPG1) and IMPG2, form a dynamic structure with distinct localization and dependency. When IMPG2 is absent, IMPG1 cannot integrate into the IPM, leading to abnormal proteoglycan accumulation and visual deficits. This work adds a new layer of understanding to IPM physiology and describes the pathologic events following deficits in proteoglycans, providing novel possibilities for visual restoration in patients with IMPG-related pathologies.
Sigma-Aldrich
Toluidine Blue, suitable for microscopy, 8.74% (ZN (THEORY))
Sigma-Aldrich
N-Acetyl-D-galactosamine, ≥98% (HPLC)
Sigma-Aldrich
Anti-IMPG2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody