Merck
  • The impact of discharge reduction activities on the occurrence of contaminants of emerging concern in surface water from the Pearl River.
The impact of discharge reduction activities on the occurrence of contaminants of emerging concern in surface water from the Pearl River.
Environmental science and pollution research international (2020-05-29)
Peng Chen, Yi Zhong, Kuncai Chen, Chongshan Guo, Jian Gong, Dedong Wang, Yan Yang, Shengtao Ma, Yingxin Yu
ABSTRACT
"Swimming across the Pearl River" is an annual large-scale sporting event with great popularity in Guangzhou. To reduce the risk of swimmers' exposure to various contaminants in the Pearl River during swimming activities, the local government limits direct sewage and effluent discharge from urban channels during the event. However, the impact of discharge reduction on some contaminants of emerging concern (CECs), such as organophosphorus flame retardants (OPFRs), bisphenol analogues (BPs), and triclosan remains unknown. In the present study, the concentrations of CECs, as well as ammonia-nitrogen (NH3-N), dissolved organic carbon, and chemical oxygen demand, were measured in aqueous and suspended particulate matter (SPM) from the Guangzhou reaches of the Pearl River. The concentration ranges of sixteen OPFRs, eight BPs, and triclosan were 21.2-91.0, 8.46-37.3, and 1.47-5.62 ng/L, respectively, in aqueous samples, and 25.2-492, 14.0-86.3, and 0.69-17.5 ng/g, respectively, in SPM samples. Hydrophobic and π-π interactions could be contributing to the distribution of CECs. Principal component analysis indicated that consumer materials, manufacturing, and domestic sewage might be the main sources of the CECs. In addition, our study showed that the concentrations of CECs did not change considerably before or after discharge reduction activities, although NH3-N showed a substantial decrease following pollution control measure. The results demonstrated that temporary reductions of contaminant discharges to the Pearl River had only limited effect on the levels of CECs. Further research is needed to investigate the distributions and potential health risks of CECs in the Pearl River.
Sigma-Aldrich
4,4′-Cyclohexylidenebisphenol, 98%
Sigma-Aldrich
Diphenyl phosphate, 99%
Sigma-Aldrich
Tripropyl phosphate, 99%
Sigma-Aldrich
4,4′-(1-Phenylethylidene)bisphenol, 99%