Merck
  • Bile Acids Quantification by Liquid Chromatography-Tandem Mass Spectrometry: Method Validation, Reference Range, and Interference Study.
Bile Acids Quantification by Liquid Chromatography-Tandem Mass Spectrometry: Method Validation, Reference Range, and Interference Study.
Diagnostics (Basel, Switzerland) (2020-07-11)
Elisa Danese, Davide Negrini, Mairi Pucci, Simone De Nitto, Davide Ambrogi, Simone Donzelli, Patricia M-J Lievens, Gian Luca Salvagno, Giuseppe Lippi
ABSTRACT
Bile acids (BA) play a pivotal role in cholesterol metabolism. Their blood concentration has also been proposed as new prognostic and diagnostic indicator of hepatobiliary, intestinal, and cardiovascular disease. Liquid chromatography tandem mass spectrometry (LC-MS/MS) currently represents the gold standard for analysis of BA profile in biological samples. We report here development and validation of a LC-MS/MS technique for simultaneously quantifying 15 BA species in serum samples. We also established a reference range for adult healthy subjects (n = 130) and performed a preliminary evaluation of in vitro and in vivo interference. The method displayed good linearity, with high regression coefficients (>0.99) over a range of 5 ng/mL (lower limit of quantification, LLOQ) and 5000 ng/mL for all analytes tested. The accuracies were between 85-115%. Both intra- and inter-assay imprecision was <10%. The recoveries ranged between 92-110%. Each of the tested BA species (assessed on three concentrations) were stable for 15 days at room temperature, 4 °C, and -20 °C. The in vitro study did not reveal any interference from triglycerides, bilirubin, or cell-free hemoglobin. The in vivo interference study showed that pools obtained from hyper-cholesterolemic patients and hyper-bilirubinemic patients due to post-hepatic jaundice for benign cholestasis, cholangiocarcinoma and pancreatic head tumors had clearly distinct patterns of BA concentrations compared with a pool obtained from samples of healthy subjects. In conclusion, this study proposes a new suitable candidate method for identification and quantitation of BA in biological samples and provides new insight into a number of variables that should be taken into account when investigating pathophysiological changes of BA in human diseases.
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder