Merck
  • Structural insights into differences in G protein activation by family A and family B GPCRs.
Structural insights into differences in G protein activation by family A and family B GPCRs.
Science (New York, N.Y.) (2020-08-01)
Daniel Hilger, Kaavya Krishna Kumar, Hongli Hu, Mie Fabricius Pedersen, Evan S O'Brien, Lise Giehm, Christine Jennings, Gözde Eskici, Asuka Inoue, Michael Lerch, Jesper Mosolff Mathiesen, Georgios Skiniotis, Brian K Kobilka
ABSTRACT
Family B heterotrimeric guanine nucleotide-binding protein (G protein)-coupled receptors (GPCRs) play important roles in carbohydrate metabolism. Recent structures of family B GPCR-Gs protein complexes reveal a disruption in the α-helix of transmembrane segment 6 (TM6) not observed in family A GPCRs. To investigate the functional impact of this structural difference, we compared the structure and function of the glucagon receptor (GCGR; family B) with the β2 adrenergic receptor (β2AR; family A). We determined the structure of the GCGR-Gs complex by means of cryo-electron microscopy at 3.1-angstrom resolution. This structure shows the distinct break in TM6. Guanosine triphosphate (GTP) turnover, guanosine diphosphate release, GTP binding, and G protein dissociation studies revealed much slower rates for G protein activation by the GCGR compared with the β2AR. Fluorescence and double electron-electron resonance studies suggest that this difference is due to the inability of agonist alone to induce a detectable outward movement of the cytoplasmic end of TM6.
Sigma-Aldrich
2,2-Dimethyl-1,3-propanediol, 99%
Supelco
Serotonin, analytical standard
Sigma-Aldrich
ANTI-FLAG® M2-Peroxidase (HRP) antibody, Mouse monoclonal, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Serotonin hydrochloride, powder
Sigma-Aldrich
Casein solution from bovine milk, 5% in water
Sigma-Aldrich
(−)-Isoproterenol hydrochloride
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O, Molecular Biology