Merck
  • miR-205-5p inhibits thymic epithelial cell proliferation via FA2H-TFAP2A feedback regulation in age-associated thymus involution.
miR-205-5p inhibits thymic epithelial cell proliferation via FA2H-TFAP2A feedback regulation in age-associated thymus involution.
Molecular immunology (2020-05-07)
Bishuang Gong, Xintong Wang, Boning Li, Ying Li, Rui Lu, Kaizhao Zhang, Bingxin Li, Yongjiang Ma, Yugu Li
ABSTRACT
Thymic epithelial cells (TECs) are essential regulators of T cell development and selection. microRNAs (miRNAs) play critical roles in regulating TECs proliferation during thymus involution. miR-205-5p is highly expressed in TECs and increases with age. However, the function and potential mechanism of miR-205-5p in TECs are not clear. miRNA expression was profiled using TECs from male and female mice at 1 and 3 months old. A total of 325 differentially expressed miRNAs (DEMs) were detected at different ages in two sexes. 24 of the DEMs had the same trend between males and females. Among them, miR-205-5p had the highest fold change. Our results showed that the expression of miR-205-5p was dramatically increased in TECs from 1 to 9 months old mice. miR-205-5p mimic inhibited TECs proliferation. Moreover, we confirmed that Fa2h was the direct target gene of miR-205-5p and FA2H was significantly decreased in TECs with increased expression of miR-205-5p. Silencing of Fa2h inhibited TECs proliferation. Furthermore, we found that the expression of Tfap2a could be promoted by FA2H and that TFAP2A could interact with miR-205-5p in TECs. Overall, miR-205-5p is an important regulator of TECs proliferation and regulates age-associated thymus involution via the miR-205-5p-FA2H-TFAP2A feedback regulatory circuit. miR-205-5p might act as a potential biomarker in TECs for age-related thymus involution.
Sigma-Aldrich
MISSION® esiRNA, targeting human TFAP2A
Sigma-Aldrich
MISSION® esiRNA, targeting human FA2H
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pcbp1