Merck
  • Brain-Sparing Sympathofacilitators Mitigate Obesity without Adverse Cardiovascular Effects.
Brain-Sparing Sympathofacilitators Mitigate Obesity without Adverse Cardiovascular Effects.
Cell metabolism (2020-05-14)
Inês Mahú, Andreia Barateiro, Eva Rial-Pensado, Noelia Martinéz-Sánchez, Sandra H Vaz, Pedro M S D Cal, Benjamin Jenkins, Tiago Rodrigues, Carlos Cordeiro, Miguel F Costa, Raquel Mendes, Elsa Seixas, Mafalda M A Pereira, Nadiya Kubasova, Vitka Gres, Imogen Morris, Carolina Temporão, Marta Olivares, Yolanda Sanz, Albert Koulman, Francisco Corzana, Ana M Sebastião, Miguel López, Gonçalo J L Bernardes, Ana I Domingos
ABSTRACT
Anti-obesity drugs in the amphetamine (AMPH) class act in the brain to reduce appetite and increase locomotion. They are also characterized by adverse cardiovascular effects with origin that, despite absence of any in vivo evidence, is attributed to a direct sympathomimetic action in the heart. Here, we show that the cardiac side effects of AMPH originate from the brain and can be circumvented by PEGylation (PEGyAMPH) to exclude its central action. PEGyAMPH does not enter the brain and facilitates SNS activity via theβ2-adrenoceptor, protecting mice against obesity by increasing lipolysis and thermogenesis, coupled to higher heat dissipation, which acts as an energy sink to increase energy expenditure without altering food intake or locomotor activity. Thus, we provide proof-of-principle for a novel class of exclusively peripheral anti-obesity sympathofacilitators that are devoid of any cardiovascular and brain-related side effects.
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Supelco
Butoxamine hydrochloride, analytical standard, for drug analysis, mixture of diastereomers
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid
Sigma-Aldrich
Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol
Sigma-Aldrich
Glycerol Standard Solution, 2.5 mg/ml equivalent triolein concentration
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit