Merck
  • Protein L from Peptostreptococcus magnus binds to the kappa light chain variable domain.
Protein L from Peptostreptococcus magnus binds to the kappa light chain variable domain.
The Journal of biological chemistry (1992-02-05)
B H Nilson, A Solomon, L Björck, B Akerström
ABSTRACT
Protein L is an immunoglobulin light chain-binding protein expressed by some strains of the anaerobic bacterial species Peptostreptococcus magnus. The major variable region subgroups of human kappa and lambda light chains were tested for protein L binding; V kappa I, V kappa III, and V kappa IV bound protein L, whereas no binding occurred with proteins of the V kappa II subgroup or with any lambda light chain subgroups. Studies of the protein L binding capacity of naturally occurring VL fragments, and VL- and CL-related trypsin- and pepsin-derived peptides prepared from a kappa I light chain, localized the site of interaction to the VL domain. The affinity constant for the binding to an isolated V kappa I fragment was comparable to that for the native protein (Ka 0.9 x 10(9) M-1 and Ka 1.5 x 10(9) M-1, respectively). No binding occurred with CL-related fragments. Extensive reduction and alkylation of the V kappa fragment or the native kappa chain resulted in complete loss of protein L binding. Although it is possible, from comparative amino acid sequence data, to identify certain VL-framework region residues that account for the selective binding of protein L by kappa I, kappa III, and kappa IV proteins, our studies indicate that this interaction is essentially dependent upon the tertiary structural integrity of the kappa chain VL domain.