Merck
  • DDX3X Suppresses the Susceptibility of Hindbrain Lineages to Medulloblastoma.
DDX3X Suppresses the Susceptibility of Hindbrain Lineages to Medulloblastoma.
Developmental cell (2020-06-20)
Deanna M Patmore, Amir Jassim, Erica Nathan, Reuben J Gilbertson, Daniel Tahan, Nadin Hoffmann, Yiai Tong, Kyle S Smith, Thirumala-Devi Kanneganti, Hiromichi Suzuki, Michael D Taylor, Paul Northcott, Richard J Gilbertson
ABSTRACT
DEAD-Box Helicase 3 X-Linked (DDX3X) is frequently mutated in the Wingless (WNT) and Sonic hedghog (SHH) subtypes of medulloblastoma-the commonest malignant childhood brain tumor, but whether DDX3X functions as a medulloblastoma oncogene or tumor suppressor gene is not known. Here, we show that Ddx3x regulates hindbrain patterning and development by controlling Hox gene expression and cell stress signaling. In mice predisposed to Wnt- or Shh medulloblastoma, Ddx3x sensed oncogenic stress and suppressed tumor formation. WNT and SHH medulloblastomas normally arise only in the lower and upper rhombic lips, respectively. Deletion of Ddx3x removed this lineage restriction, enabling both medulloblastoma subtypes to arise in either germinal zone. Thus, DDX3X is a medulloblastoma tumor suppressor that regulates hindbrain development and restricts the competence of cell lineages to form medulloblastoma subtypes.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Cresyl Violet acetate, certified by the BSC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VIII, lyophilized powder, 300-1,000 U/mg