Merck
  • Transgenic expression of Hsc70 in pancreatic islets enhances autoimmune diabetes in response to beta cell damage.
Transgenic expression of Hsc70 in pancreatic islets enhances autoimmune diabetes in response to beta cell damage.
Journal of immunology (Baltimore, Md. : 1950) (2009-10-09)
Masih-ul Alam, Julie A Harken, Anna-Maria Knorn, Alisha R Elford, Kip Wigmore, Pamela S Ohashi, Douglas G Millar
ABSTRACT
Inflammation following tissue damage promotes lymphocyte recruitment, tissue remodeling, and wound healing while maintaining self tolerance. Endogenous signals associated with tissue damage and cell death have been proposed to initiate and instruct immune responses following injury. In this study, we have examined the effects of elevated levels of a candidate endogenous danger signal, heat shock cognate protein 70 (hsc70), on stimulation of inflammation and autoimmunity following cell damage. We find that damage to pancreatic beta cells expressing additional cytosolic hsc70 leads to an increased incidence of diabetes in a transgenic mouse model. Steady-state levels of activated APC and T cell populations in the draining lymph node were enhanced, which further increased following streptozotocin-induced beta cell death. In addition, proinflammatory serum cytokines, and lymphocyte recruitment were increased in hsc70 transgenic mice. Islet Ag-specific T cells underwent a greater extent of proliferation in the lymph nodes of mice expressing hsc70 following beta cell damage, suggesting elevated Ag presentation following release of Ag in the presence of hsc70. These findings suggest that an elevated content of hsc70 in cells undergoing necrotic or apoptotic cell death can increase the extent of sterile inflammation and increase the susceptibility to autoimmunity.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, ≥10,000 EU/mg, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, purified by gel-filtration chromatography
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type X-S, lyophilized powder, 100,000-250,000 units/g solid (without added oxygen)
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, Ready Made solution, 1 mg/mL, 0.2 μm filtered
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, purified by trichloroacetic acid extraction
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, γ-irradiated, BioXtra, suitable for cell culture